(2-Aminothiazol-4-yl)methanol ≥95%

Supplier: Matrix Scientific

071982-1G 071982-5G 071982-10G
102522-454EA 173.85 USD
102522-454 102522-456 102522-458
(2-Aminothiazol-4-yl)methanol ≥95%
(2-Aminothiazol-4-yl)methanol

Hazard: Irritant

Formula: C₄H₆N₂OS
MDL Number: MFCD08275702
CAS Number: 51307-43-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR