5-Bromo-2-((tert-butoxycarbonyl)amino)thiazole-4-carboxylic acid ≥95%

Supplier: Matrix Scientific

071978-1G 071978-5G 071978-10G
102522-430EA 355.38 USD
102522-430 102522-432 102522-434
5-Bromo-2-((tert-butoxycarbonyl)amino)thiazole-4-carboxylic acid ≥95%
5-Bromo-2-((tert-butoxycarbonyl)amino)thiazole-4-carboxylic acid

Hazard: Irritant

Formula: C₉H₁₁BrN₂O₄S
CAS Number: 914347-09-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR