3-Phenylpentan-3-amine hydrochloride ≥95%

Supplier: Matrix Scientific

071975-1G 071975-5G 071975-10G
102522-412EA 710.77 USD
102522-412 102522-414 102522-416
3-Phenylpentan-3-amine hydrochloride ≥95%
3-Phenylpentan-3-amine hydrochloride

Hazard: Irritant

Formula: C11H18ClN
CAS Number: 104177-96-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR