2-Methyl-5-nitrobenzaldehyde ≥95%

Supplier: Matrix Scientific

071973-1G 071973-5G
102522-402EA 300 USD
102522-402 102522-404
2-Methyl-5-nitrobenzaldehyde ≥95%
2-Methyl-5-nitrobenzaldehyde

Hazard: Irritant

Formula: C₈H₇NO₃
MW: 165.1500 g/mol
MDL Number: MFCD03840506
CAS Number: 16634-91-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR