Ethyl-3-amino-4-pyridinecarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 3-amino-4-pyridinecarboxylate, Ethyl 3-Aminoisonicotinate, 3-Aminoisonicotinic acid ethyl ester, Ethyl 3-Aminopyridine-4-carboxylate, 3-Aminopyridine-4-carboxylic acid ethyl ester

071971-1G 071971-5G 071971-10G
102522-390EA 41.54 USD
102522-390 102522-392 102522-394
Ethyl-3-amino-4-pyridinecarboxylate ≥95%
Ethyl-3-amino-4-pyridinecarboxylate

Hazard: Irritant

Formula: C₈H₁₀N₂O₂
MW: 166.1800 g/mol
MDL Number: MFCD03788270
CAS Number: 14208-83-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR