Methyl-4-((tert-butoxycarbonyl)amino)thiophene-3-carboxylate ≥95%

Supplier: Matrix Scientific

071970-1G 071970-5G 071970-10G
102522-384EA 427.69 USD
102522-384 102522-386 102522-388
Methyl-4-((tert-butoxycarbonyl)amino)thiophene-3-carboxylate ≥95%
Methyl-4-((tert-butoxycarbonyl)amino)thiophene-3-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₅NO₄S
CAS Number: 161940-20-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR