4-(Pyrrolidin-1-yl)aniline dihydrochloride ≥95%

Supplier: Matrix Scientific

071969-1G 071969-5G 071969-10G
102522-378EA 138.46 USD
102522-378 102522-380 102522-382
4-(Pyrrolidin-1-yl)aniline dihydrochloride ≥95%
4-(Pyrrolidin-1-yl)aniline dihydrochloride

Hazard: Irritant

Formula: C₁₀H₁₆Cl₂N₂
CAS Number: 163260-77-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR