1-Boc-4-cyanopiperidine ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 4-cyanopiperidine-1-carboxylate, N-BOC-Piperidine-4-carbonitrile

071968-5G 071968-25G 071968-100G
102522-374EA 36.92 USD
102522-374 102551-786 102551-788
1-Boc-4-cyanopiperidine ≥95%
1-Boc-4-cyanopiperidine

Hazard: Toxic

Formula: C₁₁H₁₈N₂O₂
MW: 210.27 g/mol
Melting Pt: 45…48 °C
MDL Number: MFCD01861223
CAS Number: 91419-52-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR