Methyl-1-(5-bromopyrimidin-2-yl)piperidine-4-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 1-(5-bromopyrimidin-2-yl)piperidine-4-carboxylate

071967-1G 071967-5G 071967-10G
102522-366EA 284.62 USD
102522-366 102522-368 102522-370
Methyl-1-(5-bromopyrimidin-2-yl)piperidine-4-carboxylate ≥95%
Methyl-1-(5-bromopyrimidin-2-yl)piperidine-4-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₄BrN₃O₂
MW: 300.1500 g/mol
MDL Number: MFCD08275691
CAS Number: 914347-01-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR