2-Chloro-3-methoxyphenol ≥95%
Supplier: Matrix Scientific
071966-1G
071966-5G
102522-360EA
130.77
USD
102522-360
102522-362
2-Chloro-3-methoxyphenol ≥95%
2-Chloro-3-methoxyphenol
Hazard: Irritant
Formula:
C₇H₇ClO₂ MW: 158.5823 g/mol |
MDL Number:
MFCD08275690 CAS Number: 72232-49-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...