Methyl-4-((3-methylpiperazin-1-yl)methyl)benzoate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 4-((3-methylpiperazin-1-yl)methyl)benzoate

071965-1G 071965-5G 071965-10G
102522-354EA 533.85 USD
102522-354 102522-356 102522-358
Methyl-4-((3-methylpiperazin-1-yl)methyl)benzoate ≥95%
Methyl-4-((3-methylpiperazin-1-yl)methyl)benzoate

Hazard: Irritant

Formula: C₁₄H₂₀N₂O₂
MW: 248.3200 g/mol
MDL Number: MFCD08275689
CAS Number: 914349-83-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR