Methyl-4-piperazin-1-ylmethylbenzoate ≥95%

Supplier: Matrix Scientific

071963-1G 071963-5G 071963-10G
102522-342EA 533.85 USD
102522-342 102522-344 102522-346
Methyl-4-piperazin-1-ylmethylbenzoate ≥95%
Methyl-4-piperazin-1-ylmethylbenzoate

Hazard: Irritant

Formula: C₁₃H₁₈N₂O₂
CAS Number: 86620-81-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR