2-Chlororesorcinol ≥95%

Supplier: Matrix Scientific

071955-5G 071955-10G 071955-100G
102522-294EA 355.38 USD
102522-294 102522-296 102522-298
2-Chlororesorcinol ≥95%
2-Chlororesorcinol

Hazard: Irritant

Formula: C₆H₅ClO₂
MDL Number: MFCD06658175
CAS Number: 6201-65-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR