4-Chloro-6-(pyrrolidin-1-yl)-2-(trifluoromethyl)pyrimidine ≥95%

Supplier: Matrix Scientific

071952-250MG 071952-1G 071952-5G
102522-276EA 355.38 USD
102522-276 102522-278 102522-280
4-Chloro-6-(pyrrolidin-1-yl)-2-(trifluoromethyl)pyrimidine ≥95%
4-Chloro-6-(pyrrolidin-1-yl)-2-(trifluoromethyl)pyrimidine

Hazard: Irritant

Formula: C₉H₉ClF₃N₃
CAS Number: 1189872-15-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR