6-Chloro-N,N-dimethyl-2-(trifluoromethyl)pyrimidin-4-amine ≥95%

Supplier: Matrix Scientific

071951-250MG 071951-1G 071951-5G
102522-270EA 355.38 USD
102522-270 102522-272 102522-274
6-Chloro-N,N-dimethyl-2-(trifluoromethyl)pyrimidin-4-amine ≥95%
6-Chloro-N,N-dimethyl-2-(trifluoromethyl)pyrimidin-4-amine

Hazard: Irritant

Formula: C₇H₇ClF₃N₃
MW: 225.6 g/mol
MDL Number: MFCD09743687
CAS Number: 866648-53-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR