2-Methyl-6-(trifluoromethyl)pyrimidin-4-ol ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Hydroxy-2-methyl-6-(trifluoromethyl)pyrimidine, 2-Methyl-6-(trifluoromethyl)-4-pyrimidinol

071950-1G 071950-5G 071950-10G
102522-264EA 144.62 USD
102522-264 102522-266 102522-268
2-Methyl-6-(trifluoromethyl)pyrimidin-4-ol ≥95%
2-Methyl-6-(trifluoromethyl)pyrimidin-4-ol

Hazard: Irritant

Formula: C₆H₅F₃N₂O
MW: 178.1119 g/mol
MDL Number: MFCD03265321
CAS Number: 2836-44-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR