4-Chloro-2-methyl-6-(trifluoromethyl)pyrimidine ≥95%

Supplier: Matrix Scientific

071949-1G 071949-5G
102522-260EA 640 USD
102522-260 102522-262
4-Chloro-2-methyl-6-(trifluoromethyl)pyrimidine ≥95%
4-Chloro-2-methyl-6-(trifluoromethyl)pyrimidine

Hazard: Irritant

Formula: C₆H₄ClF₃N₂
MW: 196.5576 g/mol
MDL Number: MFCD03265399
CAS Number: 5993-98-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR