6-Methyl-2-(trifluoromethyl)pyrimidin-4(3H)-one ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Methyl-2-trifluoromethylpyrimidin-4-ol

071948-1G 071948-5G
102522-256EA 355.38 USD
102522-256 102522-258
6-Methyl-2-(trifluoromethyl)pyrimidin-4(3H)-one ≥95%
6-Methyl-2-(trifluoromethyl)pyrimidin-4(3H)-one

Hazard: Irritant

Formula: C₆H₅F₃N₂O
CAS Number: 2557-79-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR