(1H-Pyrazol-1-yl)methanol ≥95%

Supplier: Matrix Scientific

071943-1G 071943-5G
102522-226EA 126.15 USD
102522-226 102522-228
(1H-Pyrazol-1-yl)methanol ≥95%
(1H-Pyrazol-1-yl)methanol

Hazard: Irritant

Formula: C₄H₆N₂O
MW: 98.1000 g/mol
MDL Number: MFCD00955584
CAS Number: 1120-82-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR