2-(7-(Benzyloxy)-1H-indol-1-yl)acetic acid ≥95%

Supplier: Matrix Scientific

071942-1G 071942-5G 071942-10G
102522-220EA 710.77 USD
102522-220 102522-222 102522-224
2-(7-(Benzyloxy)-1H-indol-1-yl)acetic acid ≥95%
2-(7-(Benzyloxy)-1H-indol-1-yl)acetic acid

Hazard: Irritant

Formula: C₁₇H₁₅NO₃
CAS Number: 914349-77-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR