Methyl-2-(2-amino-5-bromothiazol-4-yl)-2-oxoacetate ≥95%

Supplier: Matrix Scientific

071936-1G 071936-5G 071936-10G
102522-188EA 249.23 USD
102522-188 102522-190 102522-192
Methyl-2-(2-amino-5-bromothiazol-4-yl)-2-oxoacetate ≥95%
Methyl-2-(2-amino-5-bromothiazol-4-yl)-2-oxoacetate

Hazard: Irritant

Formula: C₆H₅BrN₂O₃S
CAS Number: 914349-73-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR