tert-Butyl-3-oxopyrrolidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: N-BOC-3-pyrrolidinone

071931-5G 071931-25G 071931-100G
102522-162EA 35.38 USD
102522-162 102522-164 102522-166
tert-Butyl-3-oxopyrrolidine-1-carboxylate ≥95%
tert-Butyl-3-oxopyrrolidine-1-carboxylate

Hazard: Irritant

Formula: C₉H₁₅NO₃
MW: 185.22 g/mol
Melting Pt: 34…38 °C
MDL Number: MFCD01631194
CAS Number: 101385-93-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR