(R)-N-Cbz-3,4-Dihydro-1H-isoquinolinecarboxylic acid ≥95%

Supplier: Matrix Scientific

071927-250MG 071927-1G 071927-5G
102522-140EA 498.46 USD
102522-140 102522-142 102522-144
(R)-N-Cbz-3,4-Dihydro-1H-isoquinolinecarboxylic acid ≥95%
(R)-N-Cbz-3,4-Dihydro-1H-isoquinolinecarboxylic acid

Hazard: Irritant

Formula: C₁₈H₁₇NO₄
CAS Number: 151004-88-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR