2-Methylpyrimidin-4-ol ≥95%

Supplier: Matrix Scientific

071926-1G 071926-5G 071926-10G
102522-134EA 284.62 USD
102522-134 102522-136 102522-138
2-Methylpyrimidin-4-ol ≥95%
2-Methylpyrimidin-4-ol

Hazard: Irritant

Formula: C₅H₆N₂O
MDL Number: MFCD00234103
CAS Number: 67383-35-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR