1-Bromo-2-ethoxy-4-nitrobenzene ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Bromo-5-nitrophenetole

071925-1G 071925-10G
102522-128EA 178.46 USD
102522-128 102522-132
1-Bromo-2-ethoxy-4-nitrobenzene ≥95%
1-Bromo-2-ethoxy-4-nitrobenzene

Hazard: Irritant

Formula: C₈H₈BrNO₃
MW: 246.06 g/mol
MDL Number: MFCD03790890
CAS Number: 423165-33-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR