4-(2-Thiazolyl)phenol ≥95%

Supplier: Matrix Scientific

071922-1G 071922-5G 071922-10G
102522-112EA 533.85 USD
102522-112 102522-114 102522-116
4-(2-Thiazolyl)phenol ≥95%
4-(2-Thiazolyl)phenol

Hazard: Irritant

Formula: C₉H₇NOS
MW: 177.28 g/mol
MDL Number: MFCD07368615
CAS Number: 81015-49-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR