5-Bromopyrimidine-2-carboxylic acid ≥95%

Supplier: Matrix Scientific

071919-1G 071919-5G 071919-25G
102522-096EA 109.23 USD
102522-096 102522-098 102522-100
5-Bromopyrimidine-2-carboxylic acid ≥95%
5-Bromopyrimidine-2-carboxylic acid

Hazard: Irritant

Formula: C₅H₃BrN₂O₂
MW: 203.00 g/mol
MDL Number: MFCD00496793
CAS Number: 37131-87-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR