tert-Butyl (5-bromothiazol-2-yl)carbamate ≥95%

Supplier: Matrix Scientific

071918-1G 071918-5G
102522-090EA 38.46 USD
102522-090 102522-092
tert-Butyl (5-bromothiazol-2-yl)carbamate ≥95%
tert-Butyl (5-bromothiazol-2-yl)carbamate

Hazard: Irritant

Formula: C₈H₁₁BrN₂O₂S
MW: 279.15 g/mol
MDL Number: MFCD07368614
CAS Number: 405939-39-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR