5-Methoxy-2-nitrophenylamine ≥95%

Supplier: Matrix Scientific

071915-1G 071915-5G 071915-10G
102522-076EA 23.08 USD
102522-076 102522-078 102522-080
5-Methoxy-2-nitrophenylamine ≥95%
5-Methoxy-2-nitrophenylamine

Hazard: Irritant

Formula: C7H8N2O3
Melting Pt: 125…127 °C
MDL Number: MFCD00179573
CAS Number: 16133-49-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR