Methyl-2-boc-aminothiazole-4-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 2-Boc-aminothiazole-4-carboxylate, Methyl 2-amino-1,3-thiazole-4-carboxylate, 2-BOC protected

071913-1G 071913-5G 071913-10G
102522-064EA 49.23 USD
102522-064 102522-066 102522-068
Methyl-2-boc-aminothiazole-4-carboxylate ≥95%
Methyl-2-boc-aminothiazole-4-carboxylate

Hazard: Irritant

Formula: C₁₀H₁₄N₂O₄S
MW: 258.2942 g/mol
MDL Number: MFCD06659910
CAS Number: 850429-62-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR