3-Bromoisoxazole-5-carboxylic acid ≥95%

Supplier: Matrix Scientific

071911-1G 071911-5G
102522-054EA 276.92 USD
102522-054 102522-056
3-Bromoisoxazole-5-carboxylic acid ≥95%
3-Bromoisoxazole-5-carboxylic acid

Hazard: Irritant

Formula: C4H2BrNO3
MDL Number: MFCD07368611
CAS Number: 6567-35-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR