Ethyl-3-bromoisoxazole-5-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 3-bromoisoxazole-5-carboxylate

071910-1G 071910-5G 071910-10G
102522-048EA 427.69 USD
102522-048 102522-050 102522-052
Ethyl-3-bromoisoxazole-5-carboxylate ≥95%
Ethyl-3-bromoisoxazole-5-carboxylate

Hazard: Irritant

Formula: C₆H₆BrNO₃
MW: 220.0200 g/mol
MDL Number: MFCD04035587
CAS Number: 105174-97-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR