3-Aminopyridazine hydrochloride ≥95%

Supplier: Matrix Scientific

071909-1G 071909-5G 071909-10G
102522-042EA 178.46 USD
102522-042 102522-044 102522-046
3-Aminopyridazine hydrochloride ≥95%
3-Aminopyridazine hydrochloride

Hazard: Irritant

Formula: C4H6ClN3
MDL Number: MFCD07368609
CAS Number: 89203-22-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR