2-(4-Methylpiperazin-1-ylmethyl)benzylamine ≥95%

Supplier: Matrix Scientific

Synonyms: {2-[(4-Methylpiperazin-1-yl)methyl]phenyl}methylamine

071908-1G 071908-5G 071908-10G
102522-036EA 533.85 USD
102522-036 102522-038 102522-040
2-(4-Methylpiperazin-1-ylmethyl)benzylamine ≥95%
2-(4-Methylpiperazin-1-ylmethyl)benzylamine

Hazard: Irritant

Formula: C₁₃H₂₁N₃
CAS Number: 879896-50-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR