Methyl-3-(thiazol-2-yl)benzoate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 3-(thiazol-2-yl)benzoate

071907-1G 071907-5G 071907-10G
102522-030EA 640 USD
102522-030 102522-032 102522-034
Methyl-3-(thiazol-2-yl)benzoate ≥95%
Methyl-3-(thiazol-2-yl)benzoate

Hazard: Irritant

Formula: C₁₁H₉NO₂S
MW: 219.2600 g/mol
MDL Number: MFCD08275676
CAS Number: 168618-63-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR