5-(m-Tolyl)pyridin-2-amine dihydrochloride ≥95%

Supplier: Matrix Scientific

071905-1G 071905-5G 071905-10G
102522-018EA 533.85 USD
102522-018 102522-020 102522-022
5-(m-Tolyl)pyridin-2-amine dihydrochloride ≥95%
5-(m-Tolyl)pyridin-2-amine dihydrochloride

Hazard: Irritant

Formula: C₁₂H₁₄Cl₂N₂
CAS Number: 1189458-99-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR