1-(4-Nitrophenyl)piperidine ≥95%

Supplier: Matrix Scientific

071904-5G 071904-10G
102522-014EA 35.38 USD
102522-014 102522-016
1-(4-Nitrophenyl)piperidine ≥95%
1-(4-Nitrophenyl)piperidine

Hazard: Irritant

Formula: C₁₁H₁₄N₂O₂
MW: 206.24 g/mol
Melting Pt: 114°C
MDL Number: MFCD00023662
CAS Number: 6574-15-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR