1-Ethyl-4-(4-nitrophenyl)piperazine ≥95%

Supplier: Matrix Scientific

071902-1G 071902-5G 071902-10G
102522-002EA 249.23 USD
102522-002 102522-004 102522-006
1-Ethyl-4-(4-nitrophenyl)piperazine ≥95%
1-Ethyl-4-(4-nitrophenyl)piperazine

Hazard: Irritant

Formula: C₁₂H₁₇N₃O₂
MDL Number: MFCD00614372
CAS Number: 115619-00-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR