4-(4-(Bis(4-fluorophenyl)methyl)piperazin-1-yl)aniline ≥95%

Supplier: Matrix Scientific

071900-1G 071900-5G 071900-10G
102521-990EA 284.62 USD
102521-990 102521-992 102521-994
4-(4-(Bis(4-fluorophenyl)methyl)piperazin-1-yl)aniline ≥95%
4-(4-(Bis(4-fluorophenyl)methyl)piperazin-1-yl)aniline

Hazard: Irritant

Formula: C₂₃H₂₃F₂N₃
MW: 379.4456 g/mol
MDL Number: MFCD06797796
CAS Number: 914349-65-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR