1-Benzyl-4-(4-nitrophenyl)piperazine ≥95%

Supplier: Matrix Scientific

071898-1G 071898-5G 071898-10G
102521-980EA 249.23 USD
102521-980 102521-982 102521-984
1-Benzyl-4-(4-nitrophenyl)piperazine ≥95%
1-Benzyl-4-(4-nitrophenyl)piperazine

Hazard: Irritant

Formula: C₁₇H₁₉N₃O₂
MDL Number: MFCD00215725
CAS Number: 16155-08-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR