tert-Butyl-4-(4-aminobenzyl)piperazine-1-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: 4-(4-Aminobenzyl)piperazine, N1-BOC protected, tert-Butyl 4-(4-aminobenzyl)piperazine-1-carboxylate

071897-1G 071897-5G 071897-10G
102521-974EA 640 USD
102521-974 102521-976 102521-978
tert-Butyl-4-(4-aminobenzyl)piperazine-1-carboxylate ≥95%
tert-Butyl-4-(4-aminobenzyl)piperazine-1-carboxylate

Hazard: Irritant

Formula: C₁₆H₂₅N₃O₂
MW: 291.3886 g/mol
MDL Number: MFCD06797795
CAS Number: 304897-49-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR