1-(Bis(4-fluorophenyl)methyl)-4-(4-nitrobenzyl)piperazine ≥95%

Supplier: Matrix Scientific

071895-1G 071895-5G 071895-10G
102521-962EA 284.62 USD
102521-962 102521-964 102521-966
1-(Bis(4-fluorophenyl)methyl)-4-(4-nitrobenzyl)piperazine ≥95%
1-(Bis(4-fluorophenyl)methyl)-4-(4-nitrobenzyl)piperazine

Hazard: Irritant

Formula: C₂₄H₂₃F₂N₃O₂
CAS Number: 914349-64-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR