1-Boc-4-(4-nitrobenzyl)piperazine ≥95%

Supplier: Matrix Scientific

Synonyms: 1-Boc-4-(4-Nitrobenzyl)piperazine

071894-1G 071894-5G 071894-10G
102521-956EA 427.69 USD
102521-956 102521-958 102521-960
1-Boc-4-(4-nitrobenzyl)piperazine ≥95%
1-Boc-4-(4-nitrobenzyl)piperazine

Hazard: Irritant

Formula: C₁₆H₂₃N₃O₄
MW: 321.3700 g/mol
MDL Number: MFCD03407491
CAS Number: 130636-61-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR