4-((4-(Bis(4-fluorophenyl)methyl)piperazin-1-yl)methyl)aniline ≥95%

Supplier: Matrix Scientific

071893-1G 071893-5G 071893-10G
102521-950EA 355.38 USD
102521-950 102521-952 102521-954
4-((4-(Bis(4-fluorophenyl)methyl)piperazin-1-yl)methyl)aniline ≥95%
4-((4-(Bis(4-fluorophenyl)methyl)piperazin-1-yl)methyl)aniline

Hazard: Irritant

Formula: C₂₄H₂₅F₂N₃
CAS Number: 914349-63-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR