1-Boc-4-(4-nitrophenyl)piperazine ≥95%

Supplier: Matrix Scientific

071891-1G 071891-5G 071891-10G
102521-938EA 284.62 USD
102521-938 102521-940 102521-942
1-Boc-4-(4-nitrophenyl)piperazine ≥95%
1-Boc-4-(4-nitrophenyl)piperazine

Hazard: Irritant

Formula: C₁₅H₂₁N₃O₄
MDL Number: MFCD02187326
CAS Number: 182618-86-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR