1-Benzyl-4-(4-nitrobenzyl)piperazine ≥95%
Supplier: Matrix Scientific
071889-1G
071889-5G
071889-10G
102521-926EA
284.62
USD
102521-926
102521-928
102521-930
1-Benzyl-4-(4-nitrobenzyl)piperazine ≥95%
1-Benzyl-4-(4-nitrobenzyl)piperazine
Hazard: Irritant
Formula:
C₁₈H₂₁N₃O₂ |
CAS Number:
148120-37-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...