1-Methyl-4-(4-nitrobenzyl)piperazine ≥95%
Supplier: Matrix Scientific
Synonyms:
1-(4-Nitrobenzyl)-4-methylpiperazine
071888-1G
071888-5G
071888-10G
102521-920EA
284.62
USD
102521-920
102521-922
102521-924
1-Methyl-4-(4-nitrobenzyl)piperazine ≥95%
1-Methyl-4-(4-nitrobenzyl)piperazine
Hazard: Irritant
Formula:
C₁₂H₁₇N₃O₂ |
CAS Number:
70261-81-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...