5-p-Tolylpyridin-2-ylamine ≥95%

Supplier: Matrix Scientific

Synonyms: 5-(p-Tolyl)pyridin-2-amine, 5-(4-Methylphenyl)pyridin-2-amine

071886-1G 071886-5G 071886-10G
102521-908EA 355.38 USD
102521-908 102521-910 102521-912
5-p-Tolylpyridin-2-ylamine ≥95%
5-p-Tolylpyridin-2-ylamine

Hazard: Irritant

Formula: C₁₂H₁₂N₂
MW: 184.240 g/mol
MDL Number: MFCD06797790
CAS Number: 503536-74-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR