4-Imidazol-1-yl-phenylamine ≥95%

Supplier: Matrix Scientific

Synonyms: 4-(1H-Imidazol-1-yl)aniline

071883-1G 071883-5G 071883-10G
102521-890EA 90.77 USD
102521-890 102521-892 102521-894
4-Imidazol-1-yl-phenylamine ≥95%
4-Imidazol-1-yl-phenylamine

Hazard: Irritant

Formula: C₉H₉N₃
MW: 159.1900 g/mol
MDL Number: MFCD01074865
CAS Number: 2221-00-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR