1-(4-Nitrophenyl)-1H-pyrazole ≥95%

Supplier: Matrix Scientific

071882-1G 071882-5G 071882-10G
102521-884EA 143.08 USD
102521-884 102521-886 102521-888
1-(4-Nitrophenyl)-1H-pyrazole ≥95%
1-(4-Nitrophenyl)-1H-pyrazole

Hazard: Irritant

Formula: C₉H₇N₃O₂
MW: 189.1700 g/mol
MDL Number: MFCD00187828
CAS Number: 3463-30-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR